accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7242-92-4

IUPAC Name: (1R,2R,4S)-bicyclo[2.2.1]heptan-2-amine
Synonyms: (1R,2R,4S)-bicyclo[2.2.1]heptan-2-amine
Molecular Weight (g/mol): 111.19
Molecular Formula: C7H13N
InChi Key: JEPPYVOSGKWVSJ-YUZWJPFSNA-N
SMILES: N[C@@H]1C[C@H]2CC[C@@H]1C2
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results