Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

exo-2-Aminonorbornane, 99%

Catalog No. AC176950050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC176950050 5 g Glass bottle
AC176950010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC176950050 Supplier Thermo Scientific Chemicals Supplier No. 176950050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7242-92-4 | C7H13N | 111.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7242-92-4
Molecular Formula C7H13N
Molecular Weight (g/mol) 111.19
MDL Number MFCD00078132
InChI Key JEPPYVOSGKWVSJ-YUZWJPFSNA-N
Synonym 1r,2r,4s-rel-bicyclo 2.2.1 heptan-2-amine, 1r,2r,4s-bicyclo 2.2.1 heptan-2-amine, exo-2-norbornanamine, exo-2-aminonorbornane 99, exo-2-aminonorbornane, 1alpha,4alpha-norbornane-2alpha-amine, bicyclo 2.2.1 heptan-2-amine, 1r,2r,4s-rel
PubChem CID 10240785
IUPAC Name (1S,3R,4R)-bicyclo[2.2.1]heptan-3-amine
SMILES N[C@@H]1C[C@H]2CC[C@@H]1C2

Specifications

Color Colorless to Yellow
Density 0.9300g/mL
Boiling Point 49.0°C (13.0 Mbar)
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4807
Quantity 5 g
Specific Gravity 0.93
Formula Weight 111.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material exo-2-Aminonorbornane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 230-647-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.