accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7244-78-2

IUPAC Name: 1-butoxy-4-nitrobenzene
Synonyms: P-nitrophenyl butyl ether
Molecular Weight (g/mol): 195.22
Molecular Formula: C10H13NO3
InChi Key: XCCDVVZINDJESR-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results