Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butoxy-4-nitrobenzene 99.0+%, TCI America™
SDP

Catalog No. B116225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B116225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B116225G Supplier TCI America Supplier No. B116225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7244-78-2
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.22
MDL Number MFCD00024688
InChI Key XCCDVVZINDJESR-UHFFFAOYSA-N
Synonym Butyl 4-Nitrophenyl Ether
PubChem CID 81658
IUPAC Name 1-butoxy-4-nitrobenzene
SMILES CCCCOC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Quantity 25 g
Formula Weight 195.22
Percent Purity ≥99.0% (GC)
Physical Form Crystals
Chemical Name or Material 1-Butoxy-4-nitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.