accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaCH3ONSOOO

CAS RN 7246-07-3

IUPAC Name: sodium 8-ethoxyquinoline-5-sulfonate
Synonyms: sodium 8-ethoxyquinoline-5-sulfonate
Molecular Weight (g/mol): 275.25
Molecular Formula: C11H10NNaO4S
InChi Key: KGBPIIYKHSGTAJ-UHFFFAOYSA-M
SMILES: [Na+].CCOC1=C2N=CC=CC2=C(C=C1)S([O-])(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results