accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNONO

CAS RN 7250-87-5

IUPAC Name: 1,3-bis(morpholin-4-yl)propan-2-ol
Synonyms: 1,3-bis(morpholin-4-yl)propan-2-ol
Molecular Weight (g/mol): 230.31
Molecular Formula: C11H22N2O3
InChi Key: YTAILAZJMHRYOR-UHFFFAOYSA-N
SMILES: OC(CN1CCOCC1)CN1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results