Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dimorpholyl-2-propanol, MP Biomedicals

Catalog No. ICN214707 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN214707 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN214707 Supplier MP Biomedicals, Inc Supplier No. 0521470710

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7250-87-5
Molecular Formula C11H22N2O3
Molecular Weight (g/mol) 230.308
InChI Key YTAILAZJMHRYOR-UHFFFAOYSA-N
Synonym 1,3-dimorpholinopropan-2-ol, 1,3-di morpholin-4-yl propan-2-ol, 1,3-bis morpholin-4-yl propan-2-ol, 1,3-bismorpholinopropan-2-ol, 1,3-dimorpholino-2-propanol, 1,3-di 4-morpholinyl-2-propanol, 1,3-dimorpholyl-2-propanol, 2-aminonicotinaldehyde;2-amino-3-formylpyridine
PubChem CID 81670
IUPAC Name 1,3-dimorpholin-4-ylpropan-2-ol
SMILES C1COCCN1CC(CN2CCOCC2)O

Specifications

Quantity 10 g
Chemical Name or Material 1, 3-Dimorpholyl-2-Propanol
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.