CAS RN 7251-09-4
IUPAC Name:
4-amino-2-methoxybenzonitrile
Synonyms:
4-amino-2-methoxybenzonitrile
Molecular Weight (g/mol):
148.17
Molecular Formula:
C8H8N2O
InChi Key:
KTDRJLRJAHBQDQ-UHFFFAOYSA-N
SMILES:
COC1=CC(N)=CC=C1C#N
1
–
1
of
1
results
1
4-Amino-2-methoxybenzonitrile 97.0+%, TCI America™
CAS: 7251-09-4 Molecular Formula: C8H8N2O Molecular Weight (g/mol): 148.17 MDL Number: MFCD09027303 InChI Key: KTDRJLRJAHBQDQ-UHFFFAOYSA-N Synonym: 4-Cyano-3-methoxyaniline, 5-Amino-2-cyanoanisole PubChem CID: 232735 IUPAC Name: 4-amino-2-methoxybenzonitrile SMILES: COC1=C(C=CC(=C1)N)C#N