accessibility menu, dialog, popup

UserName

Viewing profile as user:
NClFFFF

CAS RN 72537-17-8

IUPAC Name: 3-chloro-2-fluoro-5-(trifluoromethyl)pyridine
Synonyms: 3-chloro-2-fluoro-5-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 199.53
Molecular Formula: C6H2ClF4N
InChi Key: GDSROTVTTLUHCO-UHFFFAOYSA-N
SMILES: FC1=C(Cl)C=C(C=N1)C(F)(F)F
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)