Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine 98.0+%, TCI America™
SDP

Catalog No. C26715G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C26715G Supplier TCI America Supplier No. C26715G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Colorless liquid with 98.0% purity, designed for use as a laboratory reagent.

Chemical Identifiers

CAS 72537-17-8
Molecular Formula C6H2ClF4N
Molecular Weight (g/mol) 199.53
MDL Number MFCD00191918
InChI Key GDSROTVTTLUHCO-UHFFFAOYSA-N
Synonym 3-chloro-2-fluoro-5-trifluoromethyl pyridine, 2-fluoro-3-chloro-5-trifluoromethyl pyridine, 2-fluoro-3-chloro-5-trifluoromethylpyridine, pyridine, 3-chloro-2-fluoro-5-trifluoromethyl, acmc-20aied, librarion l939, 3-chloro-2-fluoro-5 trifluoromethyl pyridine, 3-chloro-2-fluoro-5-trifluoromethyl-pyridine
PubChem CID 1268075
IUPAC Name 3-chloro-2-fluoro-5-(trifluoromethyl)pyridine
SMILES FC1=C(Cl)C=C(C=N1)C(F)(F)F

Specifications

Color Colorless
Boiling Point 55°C
Quantity 5 g
UN Number 1993
Formula Weight 199.53
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.