accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 72558-82-8

IUPAC Name: 1-{[(6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-[(1-carboxy-1-methylethoxy)imino]acetamido]-2-carboxylato-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl}pyridin-1-ium
Synonyms: ceftazidime ceftazidimum ceftazidima [inn-spanish] SN 401 pentahydrate fortum fortaz tazidime in plastic container CAZ ceftazidima tazicef
Molecular Weight (g/mol): 546.57
Molecular Formula: C22H22N6O7S2
InChi Key: ORFOPKXBNMVMKC-DWVKKRMSSA-N
SMILES: CC(C)(O\N=C(/C(=O)N[C@H]1[C@H]2SCC(C[N+]3=CC=CC=C3)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results