Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ceftazidime (contains ca. 10% Na2CO3), TCI America™
SDP

Catalog No. C22255G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C22255G Supplier TCI America Supplier No. C22255G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 72558-82-8
Molecular Formula C22H22N6O7S2
Molecular Weight (g/mol) 546.573
MDL Number MFCD00072034
InChI Key ORFOPKXBNMVMKC-CEZXYXJGSA-N
Synonym CAZ
PubChem CID 133687818
IUPAC Name (6S,7S)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(2-carboxypropan-2-yloxyimino)acetyl]amino]-8-oxo-3-(pyridin-1-ium-1-ylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES CC(C)(C(=O)O)ON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)C[N+]4=CC=CC=C4)C(=O)[O-]

Specifications

Color White-Yellow
Ignition Residue 10.0 to 15.0%
Loss on Drying max. 14.0 %
Quantity 5 g
Formula Weight 546.57
Physical Form Crystalline Powder
RTECSNumber UU2225000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.