accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7256-21-5

IUPAC Name: 2-methylpiperidine; 2-methylpiperidine-1-carbodithioic acid
Synonyms: 2-methylpiperidine; 2-methylpiperidine-1-carbodithioic acid
Molecular Weight (g/mol): 274.49
Molecular Formula: C13H26N2S2
InChi Key: HIZBYGUSYOUVAB-UHFFFAOYNA-N
SMILES: CC1CCCCN1.CC1CCCCN1C(S)=S
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results