Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pipecolyldithiocarbamic Acid Pipecolinium Salt 98.0+%, TCI America™
SDP

Catalog No. P068725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P068725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P068725G Supplier TCI America Supplier No. P068725G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7256-21-5
Molecular Formula C13H26N2S2
Molecular Weight (g/mol) 274.485
MDL Number MFCD00135325
InChI Key HIZBYGUSYOUVAB-UHFFFAOYSA-N
Synonym Pipecoline Pipecolyldithiocarbamate, 2-Methyl-1-piperidinecarbodithioic Acid - 2-Methylpiperidine Complex
PubChem CID 230011
IUPAC Name 2-methylpiperidine;2-methylpiperidine-1-carbodithioic acid
SMILES CC1CCCCN1.CC1CCCCN1C(=S)S

Specifications

Melting Point 132°C
Quantity 25 g
Formula Weight 274.49
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pipecolyldithiocarbamic Acid Pipecolinium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.