accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOBr

CAS RN 72593-77-2

IUPAC Name: 1-bromo-2-[2-(2-methoxyethoxy)ethoxy]ethane
Synonyms: 1-bromo-2-[2-(2-methoxyethoxy)ethoxy]ethane
Molecular Weight (g/mol): 227.1
Molecular Formula: C7H15BrO3
InChi Key: LCCCTXULXHJDLA-UHFFFAOYSA-N
SMILES: COCCOCCOCCBr
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)