accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO

CAS RN 7262-80-8

IUPAC Name: naphthalen-2-yl tetradecanoate
Synonyms: naphthalen-2-yl tetradecanoate
Molecular Weight (g/mol): 354.53
Molecular Formula: C24H34O2
InChi Key: SKRXBZIJSNMVFA-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCC(=O)OC1=CC=C2C=CC=CC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results