Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Naphthyl Myristate 97.0+%, TCI America™
SDP

Catalog No. M078010G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M078010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M078010G Supplier TCI America Supplier No. M078010G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7262-80-8
Molecular Formula C24H34O2
Molecular Weight (g/mol) 354.534
MDL Number MFCD00037748
InChI Key SKRXBZIJSNMVFA-UHFFFAOYSA-N
Synonym Myristic Acid 2-Naphthyl Ester
PubChem CID 81684
IUPAC Name naphthalen-2-yl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OC1=CC2=CC=CC=C2C=C1

Specifications

Melting Point 67°C
Color White-Yellow
Quantity 10 g
Formula Weight 354.53
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Naphthyl Myristate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.