accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HOOOCH3

CAS RN 72657-23-9

IUPAC Name: methyl 3-hydroxy-2-methylpropanoate
Synonyms: roche ester
Molecular Weight (g/mol): 118.13
Molecular Formula: C5H10O3
InChi Key: ATCCIZURPPEVIZ-UHFFFAOYNA-N
SMILES: COC(=O)C(C)CO
Boiling Point 
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)