Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl (R)-(-)-β-hydroxyisobutyrate, 98%, Thermo Scientific Chemicals

Catalog No. AC436180050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC436180050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC436180050 Supplier Thermo Scientific Chemicals Supplier No. 436180050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 72657-23-9 | Molecular Formula: C5H10O3 | Molecular Weight: 118.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72657-23-9
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00063450
InChI Key ATCCIZURPPEVIZ-SCSAIBSYSA-N
Synonym r-methyl 3-hydroxy-2-methylpropanoate, r---3-hydroxy-2-methylpropionic acid methyl ester, methyl r---3-hydroxy-2-methylpropionate, --methyl d-beta-hydroxyisobutyrate, methyl r---3-hydroxyisobutyrate, roche ester, methyl r-3-hydroxy-2-methylpropionate, propanoic acid, 3-hydroxy-2-methyl-, methyl ester, 2r, methyl 2r-3-hydroxy-2-methylpropanoate, r---3-hydroxyisobutyric acid methyl ester
PubChem CID 5324733
IUPAC Name methyl (2R)-3-hydroxy-2-methylpropanoate
SMILES CC(CO)C(=O)OC

Specifications

Color Colorless
Density 1.0700g/mL
Boiling Point 60.0°C (4.0 Torr)
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4243
Linear Formula HOCH2CH(CH3)COOCH3
Quantity 5 g
Specific Gravity 1.07
Specific Rotation − 23.30
Solubility Information Solubility in water: insoluble. Other solubilities: in ethanol: soluble
Formula Weight 118.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl (R)-(-)-β-hydroxyisobutyrate
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.