accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOHOHCl

CAS RN 7268-35-1

IUPAC Name: 1,4-dichlorobutane-2,3-diol
Synonyms: 2,3-butanediol, 1,4-dichloro-
Molecular Weight (g/mol): 159.01
Molecular Formula: C4H8Cl2O2
InChi Key: SAUBRJOIKMVSRU-UHFFFAOYNA-N
SMILES: OC(CCl)C(O)CCl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results