CAS RN 7272-84-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Rho Kinase Inhibitor III, Rockout
The Rho Kinase Inhibitor III, Rockout, also referenced under CAS 7272-84-6, controls the biological activity of Rho Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.