Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Rho Kinase Inhibitor III, Rockout

Catalog No. 55555310MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-555-310MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-555-310MG Supplier MilliporeSigma™ Supplier No. 55555310MG
Add to Cart
Add to Cart

The Rho Kinase Inhibitor III, Rockout, also referenced under CAS 7272-84-6, controls the biological activity of Rho Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 98% by HPLC
Target Proteins ROCK
CAS 7272-84-6
Molecular Formula C13H10N2
Formula Weight 194.2g/mol
Solubility DMSO or MeOH
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.