accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 72721-02-9

IUPAC Name: 5,6-dimethoxy-1H-1,3-benzodiazole
Synonyms: 5,6-dimethoxy-1H-1,3-benzodiazole
Molecular Weight (g/mol): 178.19
Molecular Formula: C9H10N2O2
InChi Key: BTWUUHKQHOSMIN-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C2N=CNC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results