accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3O(R)NHH3N

CAS RN 72748-99-3

IUPAC Name: (2R)-1-azaniumyl-2-(methoxymethyl)pyrrolidin-1-ium
Synonyms: (2R)-1-ammonio-2-(methoxymethyl)pyrrolidin-1-ium
Molecular Weight (g/mol): 132.21
Molecular Formula: C6H16N2O
InChi Key: CJGJJXCQCKRPHC-ZCFIWIBFSA-N
SMILES: COC[C@H]1CCC[NH+]1[NH3+]
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)