Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-1-Amino-2-(methoxymethyl)pyrrolidine, 95+%

Catalog No. AC277140010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC277140010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC277140010 Supplier Thermo Scientific Chemicals Supplier No. 277140010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 72748-99-3 | C6H14N2O | 130.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72748-99-3
Molecular Formula C6H14N2O
Molecular Weight (g/mol) 130.19
MDL Number MFCD00010622
InChI Key BWSIKGOGLDNQBZ-ZCFIWIBFSA-N
Synonym r-+-1-amino-2-methoxymethyl pyrrolidine, r-2-methoxymethyl pyrrolidin-1-amine, 1-pyrrolidinamine, 2-methoxymethyl-, 2r, 2r-2-methoxymethyl pyrrolidin-1-amine, r-1-amino-2-methoxymethyl pyrrolidine, r-1-+ amino-2-methoxymethylpyrrolidine, 2-methoxymethyl-1-pyrrolidinamine #, pubchem8114, r-1-amino-2-methoxymethylpyrrolidine, 2r-2-methoxymethyl pyrrolidinylamine
PubChem CID 2724538
IUPAC Name (2R)-2-(methoxymethyl)pyrrolidin-1-amine
SMILES COCC1CCCN1N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.9700g/mL
Boiling Point 42°C (1.8 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4635 to 1.4655
Quantity 1 g
Fieser 17,15
Specific Gravity 0.97
Specific Rotation + 81.5
Formula Weight 130.19
Percent Purity 95+%
Physical Form Liquid
Chemical Name or Material (R)-(+)-1-Amino-2-(methoxymethyl)pyrrolidine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.