accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNO(Z)

CAS RN 7275-43-6

IUPAC Name: (Z)-1-oxo-1H,1'H-1λ⁵-[2,2'-bipyridinyliden]-1-ylium-1'-olate
Synonyms: (Z)-1-oxo-1H-1λ⁵-[2,2'-bipyridinyliden]-1-ylium-1'-olate
Molecular Weight (g/mol): 188.19
Molecular Formula: C10H8N2O2
InChi Key: FERMVCULDZOVOJ-KTKRTIGZSA-N
SMILES: [O-]N1C=CC=C\C1=C1/C=CC=C[N+]1=O
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)