accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3COOONH(R)(S)HOOHO(R)(S)NH(E)

CAS RN 72786-10-8

IUPAC Name: hydrogen methyl (2E)-3-{[(1S,13R,14R,17S)-4-(cyclopropylmethyl)-10,17-dihydroxy-12-oxa-4-azapentacyclo[9.6.1.0¹,¹³.0⁵,¹⁷.0⁷,¹⁸]octadeca-7,9,11(18)-trien-14-yl]carbamoyl}prop-2-enoate chloride
Synonyms: hydrogen methyl (2E)-3-{[(1S,13R,14R,17S)-4-(cyclopropylmethyl)-10,17-dihydroxy-12-oxa-4-azapentacyclo[9.6.1.0¹,¹³.0⁵,¹⁷.0⁷,¹⁸]octadeca-7,9,11(18)-trien-14-yl]carbamoyl}prop-2-enoate chloride
Molecular Weight (g/mol): 490.98
Molecular Formula: C25H31ClN2O6
InChi Key: BIPHUOBUKMPSQR-ZWZZIDKESA-N
SMILES: [H+].[Cl-].COC(=O)\C=C\C(=O)N[C@@H]1CC[C@@]2(O)C3CC4=CC=C(O)C5=C4[C@@]2(CCN3CC2CC2)[C@H]1O5
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results