accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 728035-61-8

IUPAC Name: 2-(dimethylamino)quinoline-3-carbaldehyde
Synonyms: 2-(dimethylamino)quinoline-3-carbaldehyde
Molecular Weight (g/mol): 200.24
Molecular Formula: C12H12N2O
InChi Key: RCRLBHNGTPRGJQ-UHFFFAOYSA-N
SMILES: CN(C)C1=C(C=O)C=C2C=CC=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results