Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Dimethylamino)-3-quinolinecarbaldehyde, 97+%, Thermo Scientific™

Catalog No. MO07114DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MO07114DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO07114DA Supplier Thermo Scientific Chemicals Supplier No. MO07114DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 728035-61-8
Molecular Formula C12H12N2O
Molecular Weight (g/mol) 200.24
MDL Number MFCD06010203
InChI Key RCRLBHNGTPRGJQ-UHFFFAOYSA-N
PubChem CID 1076116
IUPAC Name 2-(dimethylamino)quinoline-3-carbaldehyde
SMILES CN(C)C1=C(C=O)C=C2C=CC=CC2=N1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 2-dimethylamino quinoline-3-carbaldehyde, 2-dimethylamino-quinoline-3-carbaldehyde, 2-dimethylamino-3-quinolinecarbaldehyde, 3-quinolinecarboxaldehyde,2-dimethylamino, 2-dimethylamino-3-quinoline carbaldehyde, 2-dimethylamino quinoline-3-carboxaldehyde, 3-quinolinecarboxaldehyde, 2-dimethylamino
Formula Weight 200.24
Percent Purity 97%
Chemical Name or Material 2-(Dimethylamino)-3-quinolinecarbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.