CAS RN 73-67-6
IUPAC Name:
(4-amino-2-methylpyrimidin-5-yl)methanol
Synonyms:
toxopyrimidine
Molecular Weight (g/mol):
139.16
Molecular Formula:
C6H9N3O
InChi Key:
VUTBELPREDJDDH-UHFFFAOYSA-N
SMILES:
CC1=NC=C(CO)C(N)=N1