accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7300-34-7

H2NOONH2

CAS RN 7300-34-7

IUPAC Name: 3-[4-(3-aminopropoxy)butoxy]propan-1-amine
Synonyms: 4,9-dioxa-1,12-dodecanediamine
Molecular Weight (g/mol): 204.31
Molecular Formula: C10H24N2O2
InChi Key: YOOSAIJKYCBPFW-UHFFFAOYSA-N
SMILES: NCCCOCCCCOCCCN
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)