accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOOO

CAS RN 73003-90-4

IUPAC Name: 5-(2,5-dioxooxolan-3-yl)-7-methyl-1,3,3a,4,5,7a-hexahydro-2-benzofuran-1,3-dione
Synonyms: 6-(2,5-dioxooxolan-3-yl)-4-methyl-3a,6,7,7a-tetrahydro-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 264.23
Molecular Formula: C13H12O6
InChi Key: DGQOZCNCJKEVOA-UHFFFAOYNA-N
SMILES: CC1=CC(CC2C1C(=O)OC2=O)C1CC(=O)OC1=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)