Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(2,5-Dioxotetrahydrofuryl)-3-methyl-3-cyclohexene-1,2-dicarboxylic Anhydride 95.0+%, TCI America™
SDP

Catalog No. D1901500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1901500G 500 g
D190125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1901500G Supplier TCI America Supplier No. D1901500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 73003-90-4
Molecular Formula C13H12O6
Molecular Weight (g/mol) 264.23
MDL Number MFCD00039141
InChI Key DGQOZCNCJKEVOA-UHFFFAOYNA-N
PubChem CID 175380
IUPAC Name 5-(2,5-dioxooxolan-3-yl)-7-methyl-1,3,3a,4,5,7a-hexahydro-2-benzofuran-1,3-dione
SMILES CC1=CC(CC2C1C(=O)OC2=O)C1CC(=O)OC1=O

Specifications

Melting Point 174°C
Color White-Yellow
Quantity 500 g
Formula Weight 264.23
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-(2,5-Dioxotetrahydrofuryl)-3-methyl-3-cyclohexene-1,2-dicarboxylic Anhydride
EINECSNumber (5)-5435
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.