CAS RN 7307-04-2
IUPAC Name:
5,5-dimethylhexane-2,4-dione
Synonyms:
5,5-dimethyl-2,4-hexanedione
Molecular Weight (g/mol):
142.2
Molecular Formula:
C8H14O2
InChi Key:
LCLCVVVHIPPHCG-UHFFFAOYSA-N
SMILES:
CC(=O)CC(=O)C(C)(C)C
1
–
1
of
1
results
1