Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,5-Dimethylhexane-2,4-dione, 99%

Catalog No. AAL1310406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1310406 5 g
AAL1310403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1310406 Supplier Thermo Scientific Chemicals Supplier No. L1310406
  • $362.50
    $306.65 / Each of 1

    Save $55.85

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7307-04-2 | C9H9NO4 | 195.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7307-04-2
MDL Number MFCD00142767

Specifications

Boiling Point 60°C (10 mmHg)
Flash Point 57°C (134°F)
Molecular Formula C9H9NO4
Refractive Index 1.46
Quantity 5 g
UN Number UN1224
Beilstein 969813
Synonym methyl 3-nitro-4-methylbenzoate, 4-methyl-3-nitrobenzoic acid methyl ester, benzoic acid, 4-methyl-3-nitro-, methyl ester, methyl 3-nitro-p-toluate, 4-methyl-3-nitro-benzoic acid methyl ester, pubchem10934, acmc-1bfci, ksc497g4t, methyl4-methyl-3-nitrobenzoate, 4-methoxycarbonyl-2-nitrotoluene
InChI Key YFPBHPCMYFCRKS-UHFFFAOYSA-N
SMILES CC1=C(C=C(C=C1)C(=O)OC)[N+](=O)[O-]
IUPAC Name methyl 4-methyl-3-nitrobenzoate
Molecular Weight (g/mol) 195.17
PubChem CID 81816
Formula Weight 142.2
Percent Purity 99%
Chemical Name or Material 5,5-Dimethylhexane-2,4-dione
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 230-760-0
TSCA No

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.