accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CSOONSFClClCH3

CAS RN 731-27-1

IUPAC Name: ({[dichloro(fluoro)methyl]sulfanyl}(4-methylphenyl)sulfamoyl)dimethylamine
Synonyms: tolyfluanide
Molecular Weight (g/mol): 347.24
Molecular Formula: C10H13Cl2FN2O2S2
InChi Key: HYVWIQDYBVKITD-UHFFFAOYSA-N
SMILES: CN(C)S(=O)(=O)N(SC(F)(Cl)Cl)C1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results