accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7310-74-9

BrP(E)

CAS RN 7310-74-9

IUPAC Name: triphenyl[(2E)-3-phenylprop-2-en-1-yl]phosphanium bromide
Synonyms: triphenyl[(2E)-3-phenylprop-2-en-1-yl]phosphanium bromide
Molecular Weight (g/mol): 459.37
Molecular Formula: C27H24BrP
InChi Key: APIBROGXENTUGB-ZUQRMPMESA-M
SMILES: [Br-].C(\C=C\C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results