Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cinnamyltriphenylphosphonium Bromide 96.0+%, TCI America™
SDP

Catalog No. C128625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C128625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C128625G Supplier TCI America Supplier No. C128625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7310-74-9
Molecular Formula C27H24BrP
Molecular Weight (g/mol) 459.37
MDL Number MFCD00040767
InChI Key APIBROGXENTUGB-ZUQRMPMESA-M
PubChem CID 53471258
IUPAC Name triphenyl[(2E)-3-phenylprop-2-en-1-yl]phosphanium bromide
SMILES [Br-].C(\C=C\C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 244°C
Color White
Quantity 25 g
Formula Weight 459.37
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Cinnamyltriphenylphosphonium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.