accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7313-22-6

IUPAC Name: N,N,N',N'-tetramethylpropanediamide
Synonyms: N,N,N',N'-tetramethylpropanediamide
Molecular Weight (g/mol): 158.2
Molecular Formula: C7H14N2O2
InChi Key: AOXCXILUIVQCHH-UHFFFAOYSA-N
SMILES: CN(C)C(=O)CC(=O)N(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results