Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N',N'-Tetramethylmalonamide 97.0+%, TCI America™
SDP

Catalog No. T189125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T189125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T189125G Supplier TCI America Supplier No. T189125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7313-22-6
Molecular Formula C7H14N2O2
Molecular Weight (g/mol) 158.201
MDL Number MFCD03093641
InChI Key AOXCXILUIVQCHH-UHFFFAOYSA-N
PubChem CID 10975767
IUPAC Name N,N,N',N'-tetramethylpropanediamide
SMILES CN(C)C(=O)CC(=O)N(C)C

Specifications

Color Yellow
Boiling Point 160°C
Quantity 25 g
Formula Weight 158.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N,N′,N′-Tetramethylmalonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.