accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7314-44-5

IUPAC Name: (2,4-dimethoxyphenyl)methanol
Synonyms: (2,4-dimethoxyphenyl)methanol
Molecular Weight (g/mol): 168.19
Molecular Formula: C9H12O3
InChi Key: RNKOUSCCPHSCFE-UHFFFAOYSA-N
SMILES: COC1=CC=C(CO)C(OC)=C1
Boiling Point 
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (4)
Quantity 
  • (3)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (4)
  • (3)
  • (2)
  • (1)
  • (4)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)