Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethoxybenzyl alcohol, 98%, Thermo Scientific™

Catalog No. AAB2435822
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2435822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2435822 Supplier Thermo Scientific Chemicals Supplier No. B2435822

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2, 4-Dimethoxybenzyl alcohol is used as a medical intermediate.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store in refrigerator. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7314-44-5
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.192
MDL Number MFCD00004614
InChI Key RNKOUSCCPHSCFE-UHFFFAOYSA-N
Synonym 2,4-dimethoxybenzyl alcohol, 2,4-dimethoxyphenyl methanol, benzenemethanol, 2,4-dimethoxy, 2,4-dimethoxybenzylalcohol, 1-hydroxymethyl-2,4-dimethoxybenzene, 2,4-dimethoxyphenyl methan-1-ol, pubchem7454, acmc-209opp, 2,4-dimethoxy-benzylalcohol, 2,4-dimethoxybenzyl-alcohol
PubChem CID 81750
IUPAC Name (2,4-dimethoxyphenyl)methanol
SMILES COC1=CC(=C(C=C1)CO)OC

Specifications

Melting Point 35°C to 39°C
Boiling Point 177°C to 179°C (10 mmHg)
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 2091660
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 168.19
Percent Purity 98%
Chemical Name or Material 2,4-Dimethoxybenzyl alcohol
EINECSNumber 230-775-2
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.