accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H2C(E)

CAS RN 7319-00-8

IUPAC Name: (4E)-hexa-1,4-diene
Synonyms: trans-1,4-hexadiene
Molecular Weight (g/mol): 82.15
Molecular Formula: C6H10
InChi Key: PRBHEGAFLDMLAL-GQCTYLIASA-N
SMILES: C\C=C\CC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results