Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-1,4-Hexadiene 98.0+%, TCI America™
SDP

Catalog No. H09275ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09275ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H09275ML Supplier TCI America Supplier No. H09275ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7319-00-8
Molecular Formula C6H10
Molecular Weight (g/mol) 82.146
MDL Number MFCD00009298
InChI Key PRBHEGAFLDMLAL-GQCTYLIASA-N
PubChem CID 5365552
IUPAC Name (4E)-hexa-1,4-diene
SMILES CC=CCC=C

Specifications

Color Colorless
Quantity 5 mL
UN Number 2458
Formula Weight 82.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-1,4-Hexadiene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.