accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 732304-23-3

IUPAC Name: 6-amino-2-methyl-1,4-dihydropyrimidin-4-one hydrate
Synonyms: 6-amino-2-methyl-1H-pyrimidin-4-one hydrate
Molecular Weight (g/mol): 143.15
Molecular Formula: C5H9N3O2
InChi Key: DASZPEWSQNRDMO-UHFFFAOYSA-N
SMILES: O.CC1=NC(=O)C=C(N)N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results