accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 732306-26-2

IUPAC Name: 2-[2-bromo-5-(trifluoromethyl)phenyl]acetonitrile
Synonyms: 2-[2-bromo-5-(trifluoromethyl)phenyl]acetonitrile
Molecular Weight (g/mol): 264.05
Molecular Formula: C9H5BrF3N
InChi Key: BUPMKGVGKPGMOB-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(CC#N)=C(Br)C=C1
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results