Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-(trifluoromethyl)phenylacetonitrile, 97%, Thermo Scientific™

Catalog No. AAL1959103
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1959103 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1959103 Supplier Thermo Scientific Chemicals Supplier No. L1959103

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 732306-26-2
Boiling Point 144°C to 146°C (15mmHg)
Molecular Formula C9H5BrF3N
Quantity 1g
MDL Number MFCD04973763
UN Number UN3439
Synonym 2-2-bromo-5-trifluoromethyl phenyl acetonitrile, 2-bromo-5-trifluoromethyl phenylacetonitrile, 2-2-bromo-5-trifluoromethyl phenyl ethanenitrile, 4-bromo-3-cyanomethyl benzotrifluoride, 2-bromo-5-trifluoromethyl benzyl cyanide, 2-bromo-5-trifluoromethyl benzeneacetonitrile, benzeneacetonitrile,2-bromo-5-1,1-dimethylethyl, 2-bromo-5-trifluoromethyl phenylacetonitrile_x000d_
InChI Key BUPMKGVGKPGMOB-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1C(F)(F)F)CC#N)Br
IUPAC Name 2-[2-bromo-5-(trifluoromethyl)phenyl]acetonitrile
Molecular Weight (g/mol) 264.045
PubChem CID 2783247
Formula Weight 264.04
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-(trifluoromethyl)phenylacetonitrile
Show More Show Less
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.