accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NFF

CAS RN 732306-31-9

IUPAC Name: 3,5-difluoropyridin-2-amine
Synonyms: 3,5-difluoropyridin-2-amine
Molecular Weight (g/mol): 130.1
Molecular Formula: C5H4F2N2
InChi Key: JVLFMTZUPSBCNJ-UHFFFAOYSA-N
SMILES: NC1=NC=C(F)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results