Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3,5-difluoropyridine, 98%, Thermo Scientific Chemicals

Catalog No. AAL1972706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1972706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1972706 Supplier Thermo Scientific Chemicals Supplier No. L1972706
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 732306-31-9 | Molecular Formula: C5H4F2N2 | Molecular Weight: 130.098 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 732306-31-9
Molecular Formula C5H4F2N2
Molecular Weight (g/mol) 130.098
MDL Number MFCD03788760
InChI Key JVLFMTZUPSBCNJ-UHFFFAOYSA-N
Synonym 2-amino-3,5-difluoropyridine, 3,5-difluoro-2-pyridinamine, 2-pyridinamine, 3,5-difluoro, pubchem5550, acmc-209oq0, 3,5-difluoro-2-pyridylamine, ksc495k6p, 3,5-difluoropyridin-2-ylamine, 3,5-difluoro-pyridin-2-yl-amine, 3,5-bis fluoranyl pyridin-2-amine
PubChem CID 2783238
IUPAC Name 3,5-difluoropyridin-2-amine
SMILES C1=C(C=NC(=C1F)N)F

Specifications

Melting Point 55°C to 59°C
Quantity 5 g
UN Number UN2811
Formula Weight 130.09
Percent Purity 98%
Chemical Name or Material 2-Amino-3,5-difluoropyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.