accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 73231-34-2

IUPAC Name: 2,2-dichloro-N-[(1R,2S)-3-fluoro-1-hydroxy-1-(4-methanesulfonylphenyl)propan-2-yl]acetamide
Synonyms: florfenicol
Molecular Weight (g/mol): 358.21
Molecular Formula: C12H14Cl2FNO4S
InChi Key: AYIRNRDRBQJXIF-NXEZZACHSA-N
SMILES: CS(=O)(=O)C1=CC=C(C=C1)[C@@H](O)[C@@H](CF)NC(=O)C(Cl)Cl
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results